JWPH942F330F J & W Pharmlab LLC

3-Methyl-pyridine-2-carboxamidine

  • MDL number MFCD09751910
  • Cas Number : 742028-44-0
  • Molecular Weight : 135.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/12R0115

Safety Information

MSDS https://www.jwpharmlab.com/home/product/12R0115
Availability details for
Quantity
  • Availability
  • Add to Cart