JWPH942F3323 J & W Pharmlab LLC

2-Bromo-4-methoxy-benzamidine

  • MDL number MFCD11858317
  • Cas Number : 1123169-47-0
  • Molecular Weight : 229.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/12R0101

Safety Information

MSDS https://www.jwpharmlab.com/home/product/12R0101
Availability details for
Quantity
  • Availability
  • Add to Cart