JWPH942F3338 J & W Pharmlab LLC

2-Amino-2-ethyl-hexanoic acid

  • MDL number MFCD02662647
  • Cas Number : 114781-15-6
  • Molecular Weight : 159.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/15R0129

Safety Information

MSDS https://www.jwpharmlab.com/home/product/15R0129
Availability details for
Quantity
  • Availability
  • Add to Cart