JWPH942F3339 J & W Pharmlab LLC

(S)-Amino-naphthalen-2-yl-acetic acid

  • MDL number MFCD06858439
  • Cas Number : 93779-35-2
  • Molecular Weight : 201.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/15R0132

Safety Information

MSDS https://www.jwpharmlab.com/home/product/15R0132
Availability details for
Quantity
  • Availability
  • Add to Cart