JWPH942F3343 J & W Pharmlab LLC

(R)-2-Amino-3-(3-hydroxy-5-methyl-isoxazol-4-yl)-propionic acid

  • MDL number MFCD00672629
  • Cas Number : 83654-13-1
  • Molecular Weight : 186.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/15R0473

Safety Information

MSDS https://www.jwpharmlab.com/home/product/15R0473
Availability details for
Quantity
  • Availability
  • Add to Cart