JWPH942F3365 J & W Pharmlab LLC

1-(3-Amino-phenyl)-azetidine-3-carboxylic acid methyl ester

  • MDL number MFCD07787205
  • Cas Number : 887595-89-3
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/15R0205

Safety Information

MSDS https://www.jwpharmlab.com/home/product/15R0205
Availability details for
Quantity
  • Availability
  • Add to Cart