JWPH942F33F7 J & W Pharmlab LLC

2-(1-Amino-ethyl)-4-bromo-benzoic acid methyl ester

  • MDL number MFCD22532090
  • Cas Number : 1391040-24-6
  • Molecular Weight : 258.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/15R0480

Safety Information

MSDS https://www.jwpharmlab.com/home/product/15R0480
Availability details for
Quantity
  • Availability
  • Add to Cart