JWPH942F340B J & W Pharmlab LLC

(2S,3S)-2-Amino-3-phenyl-butyric acid ethyl ester

  • MDL number MFCD31812859
  • Cas Number : 2331212-05-4
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/15R0536

Safety Information

MSDS https://www.jwpharmlab.com/home/product/15R0536
Availability details for
Quantity
  • Availability
  • Add to Cart