JWPH942F341C J & W Pharmlab LLC

(2S,3R)-2-Amino-3-(4-hydroxy-phenyl)-butyric acid ethyl ester

  • MDL number MFCD32664101
  • Molecular Weight : 223.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/15R0594

Safety Information

MSDS https://www.jwpharmlab.com/home/product/15R0594
Availability details for
Quantity
  • Availability
  • Add to Cart