JWPH942F341E J & W Pharmlab LLC

2-Boc-amino-3-phenyl-butyric acid

  • MDL number MFCD01823263
  • Cas Number : 630424-76-9
  • Molecular Weight : 279.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/15R0596

Safety Information

MSDS https://www.jwpharmlab.com/home/product/15R0596
Availability details for
Quantity
  • Availability
  • Add to Cart