JWPH942F3420 J & W Pharmlab LLC

(2R,3S)-2-Boc-amino-3-phenyl-butyric acid

  • MDL number MFCD02259748
  • Cas Number : 321524-48-5
  • Molecular Weight : 279.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/15R0598

Safety Information

MSDS https://www.jwpharmlab.com/home/product/15R0598
Availability details for
Quantity
  • Availability
  • Add to Cart