JWPH942F3438 J & W Pharmlab LLC

[2-(3-Bromo-phenyl)-ethyl]-(4-fluoro-phenyl)-amine

  • MDL number MFCD04114498
  • Cas Number : 885270-45-1
  • Molecular Weight : 294.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0012

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0012
Availability details for
Quantity
  • Availability
  • Add to Cart