JWPH942F3447 J & W Pharmlab LLC

2-Amino-1-(2,3-dihydro-benzo[1,4]dioxin-6-yl)-ethanone

  • MDL number MFCD00991035
  • Cas Number : 245329-81-1
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0604

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0604
Availability details for
Quantity
  • Availability
  • Add to Cart