JWPH942F3464 J & W Pharmlab LLC

(4-Amino-phenyl)-(4-isopropyl-phenyl)-methanone

  • MDL number MFCD06658530
  • Cas Number : 40292-22-6
  • Molecular Weight : 239.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0259

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0259
Availability details for
Quantity
  • Availability
  • Add to Cart