JWPH942F3469 J & W Pharmlab LLC

5-Aminomethyl-furan-3-carbonitrile hydrochloride

  • MDL number MFCD06658316
  • Cas Number : 232280-80-7
  • Molecular Weight : 158.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0274S

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0274S
Availability details for
Quantity
  • Availability
  • Add to Cart