JWPH942F346B J & W Pharmlab LLC

Acridine-9-carboxylic acid (3-amino-propyl)-amide

  • MDL number MFCD06658327
  • Cas Number : 259221-98-2
  • Molecular Weight : 279.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0281

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0281
Availability details for
Quantity
  • Availability
  • Add to Cart