JWPH942F346E J & W Pharmlab LLC

2-Amino-N-cyclopropyl-acetamide

  • MDL number MFCD06658308
  • Cas Number : 120436-02-4
  • Molecular Weight : 114.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0284

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0284
Availability details for
Quantity
  • Availability
  • Add to Cart