JWPH942F3492 J & W Pharmlab LLC

1-(3-Aminomethyl-phenyl)-2-(4-fluoro-phenyl)-ethanone

  • MDL number MFCD09997771
  • Cas Number : 1017781-75-7
  • Molecular Weight : 243.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0513

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0513
Availability details for
Quantity
  • Availability
  • Add to Cart