JWPH942F3499 J & W Pharmlab LLC

2,2,2-Trifluoro-1-thiophen-2-ylmethyl-ethylamine

  • MDL number MFCD09997779
  • Cas Number : 1012884-80-8
  • Molecular Weight : 195.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0519

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0519
Availability details for
Quantity
  • Availability
  • Add to Cart