JWPH942F34B4 J & W Pharmlab LLC

1-(4-Aminomethyl-phenyl)-2-phenyl-ethanone hydrochloride

  • MDL number MFCD09997763
  • Cas Number : 24244-39-1
  • Molecular Weight : 261.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0507S

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0507S
Availability details for
Quantity
  • Availability
  • Add to Cart