JWPH942F34C3 J & W Pharmlab LLC

[2-(2,3-Dihydro-benzo[1,4]dioxin-6-yl)-2-oxo-ethyl]-carbamic acid tert-butyl ester

  • MDL number MFCD11506030
  • Cas Number : 1086392-10-0
  • Molecular Weight : 293.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0589

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0589
Availability details for
Quantity
  • Availability
  • Add to Cart