JWPH942F34F7 J & W Pharmlab LLC

(4-Acetyl-phenyl)-methyl-carbamic acid tert-butyl ester

  • MDL number MFCD09261161
  • Cas Number : 907209-80-7
  • Molecular Weight : 249.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0712

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0712
Availability details for
Quantity
  • Availability
  • Add to Cart