JWPH942F34F9 J & W Pharmlab LLC

(4-Aminomethyl-phenyl)-methyl-carbamic acid tert-butyl ester

  • MDL number MFCD09953964
  • Cas Number : 191871-91-7
  • Molecular Weight : 236.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0715

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0715
Availability details for
Quantity
  • Availability
  • Add to Cart