JWPH942F3507 J & W Pharmlab LLC

3-(1,4,5,6-Tetrahydro-pyrimidin-2-ylsulfanyl)-propylamine

  • MDL number MFCD03042452
  • Cas Number : 7391-14-2
  • Molecular Weight : 173.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0761

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0761
Availability details for
Quantity
  • Availability
  • Add to Cart