JWPH942F3511 J & W Pharmlab LLC

(R)-N-(2-Hydroxy-2-methyl-1-phenyl-propyl)-4-methyl-benzenesulfonamide

  • MDL number MFCD09025333
  • Cas Number : 620627-46-5
  • Molecular Weight : 319.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0812

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0812
Availability details for
Quantity
  • Availability
  • Add to Cart