JWPH942F3535 J & W Pharmlab LLC

Methyl-(3-methyl-indan-1-yl)-amine

  • MDL number MFCD15474876
  • Cas Number : 876504-45-9
  • Molecular Weight : 161.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0846

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0846
Availability details for
Quantity
  • Availability
  • Add to Cart