JWPH942F3565 J & W Pharmlab LLC

tert-Butyl N-[(1R)-1-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-1-(phenylsulfonyl)methyl]carbamate

  • MDL number MFCD18914325
  • Cas Number : 657404-76-7
  • Molecular Weight : 371.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0998

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0998
Availability details for
Quantity
  • Availability
  • Add to Cart