JWPH942F3577 J & W Pharmlab LLC

2-Bromo-4-tert-butyl-6-nitro-phenylamine

  • MDL number MFCD11845936
  • Cas Number : 1027356-55-3
  • Molecular Weight : 273.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R1078

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R1078
Availability details for
Quantity
  • Availability
  • Add to Cart