JWPH942F3594 J & W Pharmlab LLC

(R)-{3-[2-(2-Hydroxy-1-methyl-ethoxy)-phenyl]-propyl}-carbamic acid benzyl ester

  • MDL number MFCD24038848
  • Cas Number : 874336-15-9
  • Molecular Weight : 343.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R1483

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R1483
Availability details for
Quantity
  • Availability
  • Add to Cart