JWPH942F35A0 J & W Pharmlab LLC

cis-4-Pyrrolidin-1-yl-cyclohexanol

  • MDL number MFCD30181884
  • Cas Number : 1965309-00-5
  • Molecular Weight : 169.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R1602

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R1602
Availability details for
Quantity
  • Availability
  • Add to Cart