JWPH942F35A5 J & W Pharmlab LLC

(2S)-2-Amino-2-(3-chloro-5-fluoro-phenyl)-ethanol

  • MDL number MFCD09254249
  • Cas Number : 1213574-69-6
  • Molecular Weight : 189.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R1607

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R1607
Availability details for
Quantity
  • Availability
  • Add to Cart