JWPH942F35A8 J & W Pharmlab LLC

N3,N3-Dimethyl-6-(trifluoromethyl)benzene-1,3-diamine

  • MDL number MFCD24368594
  • Cas Number : 1369906-40-0
  • Molecular Weight : 204.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R1356

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R1356
Availability details for
Quantity
  • Availability
  • Add to Cart