JWPH942F35AD J & W Pharmlab LLC

2,2,2-Trifluoro-1-(2-trifluoromethyl-phenyl)-ethylamine

  • MDL number MFCD07384568
  • Cas Number : 1211507-43-5
  • Molecular Weight : 243.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R1364

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R1364
Availability details for
Quantity
  • Availability
  • Add to Cart