JWPH942F35C8 J & W Pharmlab LLC

(3-Bromo-naphthalen-1-yl)-carbamic acid tert-butyl ester

  • MDL number MFCD30181882
  • Cas Number : 1965310-45-5
  • Molecular Weight : 322.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R1565

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R1565
Availability details for
Quantity
  • Availability
  • Add to Cart