JWPH942F35FF J & W Pharmlab LLC

(R)-5-Chloro-1,2,3,4-tetrahydro-naphthalen-1-ylamine

  • MDL number MFCD07373994
  • Cas Number : 1213589-52-6
  • Molecular Weight : 181.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R1515

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R1515
Availability details for
Quantity
  • Availability
  • Add to Cart