JWPH942F3613 J & W Pharmlab LLC

(1S,3S)-2-(3-Hydroxy-cyclohexyl)-isoindole-1,3-dione

  • MDL number MFCD31693555
  • Cas Number : 2381513-11-5
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R1727

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R1727
Availability details for
Quantity
  • Availability
  • Add to Cart