JWPH942F363F J & W Pharmlab LLC

4-Amino-dihydro-furan-2-one

  • MDL number MFCD09036146
  • Cas Number : 16504-58-8
  • Molecular Weight : 101.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R1590

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R1590
Availability details for
Quantity
  • Availability
  • Add to Cart