JWPH942F364D J & W Pharmlab LLC

(2R)-2-Amino-2-(4-trifluoromethyl-phenyl)-ethanol

  • MDL number MFCD09253775
  • Cas Number : 306281-86-7
  • Molecular Weight : 205.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R1621

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R1621
Availability details for
Quantity
  • Availability
  • Add to Cart