JWPH942F3665 J & W Pharmlab LLC

trans-(3-Hydroxy-cyclohexyl)-carbamic acid tert-butyl ester

  • MDL number MFCD14155670
  • Cas Number : 1425253-99-1
  • Molecular Weight : 215.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R1672

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R1672
Availability details for
Quantity
  • Availability
  • Add to Cart