JWPH942F3679 J & W Pharmlab LLC

(R)-[1-(4-Bromo-2-methyl-phenyl)-ethyl]-carbamic acid tert-butyl ester

  • MDL number MFCD31812867
  • Cas Number : 2387559-64-8
  • Molecular Weight : 314.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R1730

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R1730
Availability details for
Quantity
  • Availability
  • Add to Cart