JWPH942F36AB J & W Pharmlab LLC

1-Benzyl-azetidine-2-carboxylic acid amide

  • MDL number MFCD06658337
  • Cas Number : 40432-40-4
  • Molecular Weight : 190.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/25R0009

Safety Information

MSDS https://www.jwpharmlab.com/home/product/25R0009
Availability details for
Quantity
  • Availability
  • Add to Cart