JWPH942F36AD J & W Pharmlab LLC

(R)-2-(1-Benzyl-azetidin-2-yl)-ethanol

  • MDL number MFCD09999619
  • Cas Number : 1414960-57-8
  • Molecular Weight : 191.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/25R0085

Safety Information

MSDS https://www.jwpharmlab.com/home/product/25R0085
Availability details for
Quantity
  • Availability
  • Add to Cart