JWPH942F36B2 J & W Pharmlab LLC

2-Amino-cycloheptanol

  • MDL number MFCD09836129
  • Cas Number : 89854-90-0
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R1878

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R1878
Availability details for
Quantity
  • Availability
  • Add to Cart