JWPH942F36BC J & W Pharmlab LLC

2-Azetidin-3-yl-pyridine

  • MDL number MFCD10000579
  • Cas Number : 687993-73-3
  • Molecular Weight : 134.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/25R0134

Safety Information

MSDS https://www.jwpharmlab.com/home/product/25R0134
Availability details for
Quantity
  • Availability
  • Add to Cart