JWPH942F36C7 J & W Pharmlab LLC

(1S,2R)-2-(4-Chloro-phenyl)-cyclopropylamine

  • MDL number MFCD20699487
  • Cas Number : 209738-24-9
  • Molecular Weight : 167.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R1915

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R1915
Availability details for
Quantity
  • Availability
  • Add to Cart