JWPH942F36DE J & W Pharmlab LLC

7-Bromo-1H-indole-2-boronic acid pinacol ester

  • MDL number MFCD11858379
  • Cas Number : 1072812-23-7
  • Molecular Weight : 322.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/32R0062P

Safety Information

MSDS https://www.jwpharmlab.com/home/product/32R0062P
Availability details for
Quantity
  • Availability
  • Add to Cart