JWPH942F3702 J & W Pharmlab LLC

3-Ethylsulfonylphenylboronic acid pinacol ester

  • MDL number MFCD16660312
  • Cas Number : 2096331-73-4
  • Molecular Weight : 296.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/32R0119P

Safety Information

MSDS https://www.jwpharmlab.com/home/product/32R0119P
Availability details for
Quantity
  • Availability
  • Add to Cart