JWPH942F373C J & W Pharmlab LLC

Ethyl 6-cyano-1H-indazole-3-carboxylate

  • MDL number MFCD07371601
  • Cas Number : 885279-19-6
  • Molecular Weight : 215.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/45R0208

Safety Information

MSDS https://www.jwpharmlab.com/home/product/45R0208
Availability details for
Quantity
  • Availability
  • Add to Cart