JWPH942F3741 J & W Pharmlab LLC

6-(3,6-Dihydro-2H-thiopyran-4-yl)-1H-indazole

  • MDL number MFCD04114670
  • Cas Number : 885272-24-2
  • Molecular Weight : 216.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/45R0016

Safety Information

MSDS https://www.jwpharmlab.com/home/product/45R0016
Availability details for
Quantity
  • Availability
  • Add to Cart