JWPH942F3748 J & W Pharmlab LLC

8-Methoxy-chroman-3-carboxylic acid ethyl ester

  • MDL number MFCD04114624
  • Cas Number : 221185-47-3
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/33R0065

Safety Information

MSDS https://www.jwpharmlab.com/home/product/33R0065
Availability details for
Quantity
  • Availability
  • Add to Cart